1-Boc-4-iodopiperidine ≥97.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
1-BOC-4-iodo-piperidine, tert-Butyl 4-iodopiperidine-1-carboxylate
B4164-1G
B4164-5G
TCIAB4164-1GEA
41.8
EUR
TCIAB4164-1G
TCIAB4164-5G
1-Boc-4-iodopiperidine ≥97.0% (by GC)
1-Boc-4-iodopiperidine
Formule:
C₁₀H₁₈INO₂ Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD04115041 CAS nummer: 301673-14-3 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 44.0 to 48.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...