O-tert-Butyl-N,N'-diisopropylisourea ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: tert-Butyl N,N'-diisopropylcarbamimidate, 2-tert-Butyl-1,3-diisopropylisourea

B4178-5G B4178-1G
TCIAB4178-5GEA 171 EUR
TCIAB4178-5G TCIAB4178-1G
O-tert-Butyl-N,N'-diisopropylisourea ≥98.0% (by titrimetric analysis)
O-tert-Butyl-N,N'-diisopropylisourea
Formule: C₁₁H₂₄N₂O
MW: 200.33 g/mol
Kookpunt: 61°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD06657672
CAS nummer: 71432-55-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d87fa61f75f68c240e3a804da372a1f25fce1535&vid=d87fa61f75f68c240e3a804da372a1f25fce1535&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR