3-Bromo-4-(trifluoromethoxy)aniline ≥97.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

B4190-1G B4190-5G
TCIAB4190-1GEA 28.6 EUR
TCIAB4190-1G TCIAB4190-5G
3-Bromo-4-(trifluoromethoxy)aniline ≥97.0% (by GC, titration analysis)
3-Bromo-4-(trifluoromethoxy)aniline
Formule: C₇H₅BrF₃NO
MDL Nummer: MFCD00190127
CAS nummer: 191602-54-7

Order Now

Specification Test Results

Appearance White to Almost white clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR