5-Bromo-2-nitrobenzaldehyde ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
B4192-1G
B4192-5G
TCIAB4192-1GEA
18.7
EUR
TCIAB4192-1G
TCIAB4192-5G
5-Bromo-2-nitrobenzaldehyde ≥98.0% (by GC)
5-Bromo-2-nitrobenzaldehyde
Formule:
C₇H₄BrNO₃ |
MDL Nummer:
MFCD00456508 CAS nummer: 20357-20-4 |
Specification Test Results
Appearance | White to Light red to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 71.0 to 75.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...