1-Bromo-2,4-bis(trifluoromethyl)benzene ≥97.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
2,4-Bis(trifluoromethyl)-1-bromobenzene
B4207-1G
B4207-5G
TCIAB4207-1GEA
24.2
EUR
TCIAB4207-1G
TCIAB4207-5G
1-Bromo-2,4-bis(trifluoromethyl)benzene ≥97.0% (by GC)
1-Bromo-2,4-bis(trifluoromethyl)benzene
Formule:
C₈H₃BrF₆ Kookpunt: 51°C Vlampunt: 69°C |
MDL Nummer:
MFCD00074904 CAS nummer: 327-75-3 |
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...