Brinzolamide ≥98.0% (by HPLC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
B4258-25MG
B4258-100MG
TCIAB4258-25MGEA
29.7
EUR
TCIAB4258-25MG
TCIAB4258-100MG
Brinzolamide ≥98.0% (by HPLC)
Brinzolamide
Formule:
C₁₂H₂₁N₃O₅S₃ Bewaar temperatuur: Vriezer |
MDL Nummer:
MFCD08067749 CAS nummer: 138890-62-7 Merck Index: 14,01376 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Elemental analysis (Nitrogen) | 10.60 to 11.20 % |
Melting point | 130.0 to 134.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...