Bicyclo[2.2.2]oct-7-ene-2,3,5,6-tetracarboxylic dianhydride ≥98.0% (by GC, titration analysis), purified by sublimation
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
B4261-5G
B4261-25G
TCIAB4261-5GEA
68.2
EUR
TCIAB4261-5G
TCIAB4261-25G
Bicyclo[2.2.2]oct-7-ene-2,3,5,6-tetracarboxylic dianhydride ≥98.0% (by GC, titration analysis), purified by sublimation
Bicyclo[2.2.2]oct-7-ene-2,3,5,6-tetracarboxylic dianhydride
Formule:
C₁₂H₈O₆ MW: 248.19 g/mol Smeltpunt: 382°C |
MDL Nummer:
MFCD00082228 CAS nummer: 1719-83-1 EINECS: 217-009-2 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Methanolysis Method) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...