Methyl palmitate, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Hexadecylic acid methyl ester, Methyl hexadecanoate, Palmitic acid methyl ester, Hexadecanoic acid methyl ester, Palmitic Acid Methyl Ester
M-Clarity™ quality level = MQ100
76159-1G 76159-5G
76159-1GEA 64.3 EUR
76159-1G 76159-5G
Methyl palmitate, Supelco®
Methyl palmitate
Formule: C₁₇H₃₄O₂
MW: 270.46 g/mol
Kookpunt: 417
Smeltpunt: 29.9
Dichtheid: 0.8804 g/cm³ (20 °C)
MDL Nummer: MFCD00008994
CAS nummer: 112-39-0
EINECS: 203-966-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=07e2fce1a4cc76b91e7559abfc791ae541b93477&vid=aa6330e4046171e0dcd7616969ad0cdf4e0ec81d&prid=web&clid=SAPDAM