2-((tert-Butoxycarbonyl)amino)propanoic acid ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: N-Boc-DL-alanine, N-(tert-Butoxycarbonyl)-DL-alanine, Boc-DL-Ala-OH

B4282-5G B4282-25G
TCIAB4282-5GEA 18.7 EUR
TCIAB4282-5G TCIAB4282-25G
2-((tert-Butoxycarbonyl)amino)propanoic acid ≥98.0% (by HPLC, titration analysis)
2-((tert-Butoxycarbonyl)amino)propanoic acid
Formule: C₈H₁₅NO₄
MDL Nummer: MFCD00063124
CAS nummer: 3744-87-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 110.0 to 114.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR