tert-Butyl-3-iodoazetidine-1-carboxylate ≥95.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
B4288-5G B4288-1G
TCIAB4288-5GEA 94.6 EUR
TCIAB4288-5G TCIAB4288-1G
tert-Butyl-3-iodoazetidine-1-carboxylate ≥95.0% (by GC)
tert-Butyl-3-iodoazetidine-1-carboxylate
Formule: C₈H₁₄INO₂
MDL Nummer: MFCD09752821
CAS nummer: 254454-54-1
Order Now
Specification Test Results
Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 95.0 %