(4-Boc-aminophenyl)boronic acid (contains varying amounts of Anhydride)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

B4335-1G B4335-5G
TCIAB4335-1GEA 46.2 EUR
TCIAB4335-1G TCIAB4335-5G
(4-Boc-aminophenyl)boronic acid (contains varying amounts of Anhydride)
(4-Boc-aminophenyl)boronic acid
Formule: C₁₁H₁₆BNO₄
MDL Nummer: MFCD02093054
CAS nummer: 380430-49-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 109.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR