2-Bromo-7-iodofluorene ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
B4431-5G
B4431-25G
TCIAB4431-5GEA
47.3
EUR
TCIAB4431-5G
TCIAB4431-25G
2-Bromo-7-iodofluorene ≥98.0% (by GC)
2-Bromo-7-iodofluorene
Formule:
C₁₃H₈BrI |
MDL Nummer:
MFCD12024276 CAS nummer: 123348-27-6 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 182.0 to 186.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...