TDZD-8 ≥98.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 4-Benzyl-2-methyl-1,2,4-thiadiazolidine-3,5-dione

B4436-5MG B4436-25MG
TCIAB4436-5MGEA 85.8 EUR
TCIAB4436-5MG TCIAB4436-25MG
TDZD-8 ≥98.0% (by HPLC)
TDZD-8
Formule: C₁₀H₁₀N₂SO₂
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD04973552
CAS nummer: 327036-89-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd96accc6f46c4c4d58a87788de2e27ecc1d2334&vid=fd96accc6f46c4c4d58a87788de2e27ecc1d2334&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 12.00 to 13.00 %
Melting point 61.0 to 65.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR