2-Bromo-1,10-phenanthroline ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

B4456-1G B4456-5G
TCIAB4456-1GEA 102 EUR
TCIAB4456-1G TCIAB4456-5G
2-Bromo-1,10-phenanthroline ≥98.0% (by titrimetric analysis)
2-Bromo-1,10-phenanthroline
Formule: C₁₂H₇BrN₂
MDL Nummer: MFCD08276287
CAS nummer: 22426-14-8

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 161.0 to 165.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR