2-(4-Biphenylylcarbonyl)benzoic acid ≥97.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: o-(4-Biphenylylcarbonyl)benzoic acid

B4532-1G B4532-5G
TCIAB4532-1GEA 30.8 EUR
TCIAB4532-1G TCIAB4532-5G
2-(4-Biphenylylcarbonyl)benzoic acid ≥97.0% (by GC, titration analysis)
2-(4-Biphenylylcarbonyl)benzoic acid
Formule: C₂₀H₁₄O₃
MW: 302.33 g/mol
MDL Nummer: MFCD00392368
CAS nummer: 42797-18-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 227.0 to 231.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR