Benzo[g]quinoxaline ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

B4540-200MG B4540-1G
TCIAB4540-200MGEA 53.9 EUR
TCIAB4540-200MG TCIAB4540-1G
Benzo[g]quinoxaline ≥98.0% (by GC)
Benzo[g]quinoxaline
Formule: C₁₂H₈N₂
MDL Nummer: MFCD18449114
CAS nummer: 260-50-4

Order Now

Specification Test Results

Purity (GC) min. 98.0 %
Melting point 125.0 to 129.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR