3-Bromo-2-thenoic acid ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
B4605-1G
B4605-5G
TCIAB4605-1GEA
17.6
EUR
TCIAB4605-1G
TCIAB4605-5G
3-Bromo-2-thenoic acid ≥98.0% (by GC, titration analysis)
3-Bromo-2-thenoic acid
Formule:
C₅H₃BrO₂S |
MDL Nummer:
MFCD00052291 CAS nummer: 7311-64-0 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 196.0 to 200.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...