Benzyl acrylate ≥97.0% (by GC) stabilised
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
Acrylic Acid Benzyl Ester
Stabilisatie:
Stabilised with 4-methoxyphenol (MEHQ)
B4627-25G
B4627-100G
TCIAB4627-25GEA
51.7
EUR
TCIAB4627-25G
TCIAB4627-100G
Benzyl acrylate ≥97.0% (by GC) stabilised
Benzyl acrylate
Formule:
C₁₀H₁₀O₂ MW: 162.19 g/mol Kookpunt: 60°C Vlampunt: 108°C |
MDL Nummer:
MFCD00048147 CAS nummer: 2495-35-4 EINECS: 219-673-9 |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...