3-Bromo-1H-pyrazolo[3,4-d]pyrimidin-4-amine ≥97.0% (by HPLC, total nitrogen)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

B4630-1G B4630-5G
TCIAB4630-1GEA 17.6 EUR
TCIAB4630-1G TCIAB4630-5G
3-Bromo-1H-pyrazolo[3,4-d]pyrimidin-4-amine ≥97.0% (by HPLC, total nitrogen)
3-Bromo-1H-pyrazolo[3,4-d]pyrimidin-4-amine
Formule: C5H4BrN5
Smeltpunt: 350°C
MDL Nummer: MFCD12196922
CAS nummer: 83255-86-1

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR