(5-bromothien-2-yl)methanol ≥97.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
(5-Bromothien-2-yl)methanol, 5-Bromo-2-thiophenemethanol
B4651-1G
B4651-5G
TCIAB4651-1GEA
60.5
EUR
TCIAB4651-1G
TCIAB4651-5G
(5-bromothien-2-yl)methanol ≥97.0% (by GC)
(5-bromothien-2-yl)methanol
Formule:
C₅H₅BrOS |
MDL Nummer:
MFCD06202626 CAS nummer: 79387-71-6 |
Specification Test Results
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...