3-Bromo-4-fluorobenzyl alcohol 98,0 GC_ASSAY_METHOD
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: (3-Bromo-4-fluorophenyl)methanol
B4656-5G B4656-25G
TCIAB4656-5GEA 34.1 EUR
TCIAB4656-5G TCIAB4656-25G
3-Bromo-4-fluorobenzyl alcohol 98,0 GC_ASSAY_METHOD
3-Bromo-4-fluorobenzyl alcohol
Formule: C₇H₆BrFO
MW: 205.03
Kookpunt: 90°C
MDL Nummer: MFCD00143093
CAS nummer: 77771-03-0
Order Now
Specification Test Results
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %