N,N-Bis(4-bromophenyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 4-[Bis(4-bromophenyl)amino]phenylboronic acid Pinacol ester, 2-[4-[Bis(4-bromophenyl)amino]phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-Bromo-N-(4-bromophenyl)-N-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)aniline

B4660-200MG B4660-1G
TCIAB4660-200MGEA 149 EUR
TCIAB4660-200MG TCIAB4660-1G
N,N-Bis(4-bromophenyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline ≥98.0% (by HPLC, titration analysis)
N,N-Bis(4-bromophenyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Formule: C₂₄H₂₄BBr₂NO₂
MW: 529.08 g/mol
MDL Nummer: MFCD30721938
CAS nummer: 850153-24-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=042e327331e828fccc0ca2a729c4b9168ca72276&vid=042e327331e828fccc0ca2a729c4b9168ca72276&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 146.0 to 150.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR