Bis(2,2,2-trifluoroethyl) Carbonate ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Carbonic acid bis(2,2,2-trifluoroethyl) ester
B4703-5G
B4703-25G
TCIAB4703-5GEA
67.1
EUR
TCIAB4703-5G
TCIAB4703-25G
Bis(2,2,2-trifluoroethyl) Carbonate ≥98.0% (by GC)
Bis(2,2,2-trifluoroethyl) Carbonate
Formule:
C₅H₄F₆O₃ Kookpunt: 118°C |
MDL Nummer:
MFCD11857845 CAS nummer: 1513-87-7 UN: 3272 ADR: 3,II |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...