4-Bromo-1,3,5-trimethyl-1H-pyrazole ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 4-Bromo-1,3,5-trimethylpyrazole

B4744-1G B4744-5G
TCIAB4744-1GEA 23.1 EUR
TCIAB4744-1G TCIAB4744-5G
4-Bromo-1,3,5-trimethyl-1H-pyrazole ≥98.0% (by GC, titration analysis)
4-Bromo-1,3,5-trimethyl-1H-pyrazole
Formule: C₆H₉BrN₂
Kookpunt: 97°C
MDL Nummer: MFCD00052532
CAS nummer: 15801-69-1

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR