3-Bromo-2,6-dimethoxytoluene ≥97.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
B4745-200MG
B4745-1G
TCIAB4745-200MGEA
42.9
EUR
TCIAB4745-200MG
TCIAB4745-1G
3-Bromo-2,6-dimethoxytoluene ≥97.0% (by GC)
3-Bromo-2,6-dimethoxytoluene
Formule:
C₉H₁₁BrO₂ Kookpunt: 96°C |
MDL Nummer:
MFCD11046750 CAS nummer: 22794-95-2 |
Specification Test Results
Appearance | Light yellow to Yellow to Orange clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...