2-Bromo-4-fluoropyridine ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
B4760-1G
B4760-5G
TCIAB4760-1GEA
50.6
EUR
TCIAB4760-1G
TCIAB4760-5G
2-Bromo-4-fluoropyridine ≥98.0% (by GC)
2-Bromo-4-fluoropyridine
Formule:
C₅H₃BrFN Vlampunt: 76°C |
MDL Nummer:
MFCD09260909 CAS nummer: 357927-50-5 |
Specification Test Results
Appearance | White to Almost white clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...