tert-Butyl-3-cyano-4-oxopyrrolidine-1-carboxylate ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
3-Cyano-4-oxopyrrolidine-1-carboxylic acid tert-Butyl ester, tert-Butyl 3-cyano-4-oxopyrrolidine-1-carboxylate, 1-Boc-3-cyano-4-pyrrolidone, tert-Butyl 3-Cyano-4-oxopyrrolidine-1-carboxylate, 1-tert-Butoxycarbonyl-3-cyano-4-pyrrolidone, 1-Boc-3-cyano-4-pyrrolidinone, 1-tert-Butoxycarbonyl-3-cyano-4-pyrrolidinone
B4761-1G
B4761-5G
TCIAB4761-1GEA
30.8
EUR
TCIAB4761-1G
TCIAB4761-5G
tert-Butyl-3-cyano-4-oxopyrrolidine-1-carboxylate ≥98.0% (by GC)
tert-Butyl-3-cyano-4-oxopyrrolidine-1-carboxylate
Formule:
C₁₀H₁₄N₂O₃ MW: 210.23 g/mol Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD01318456 CAS nummer: 175463-32-8 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...