ω-Bromo-2,4-dimethoxyacetophenone ≥97.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
B4768-1G
B4768-5G
TCIAB4768-1GEA
157
EUR
TCIAB4768-1G
TCIAB4768-5G
ω-Bromo-2,4-dimethoxyacetophenone ≥97.0% (by GC)
ω-Bromo-2,4-dimethoxyacetophenone
Formule:
C₁₀H₁₁BrO₃ |
MDL Nummer:
MFCD00000197 CAS nummer: 60965-26-6 EINECS: 262-542-6 UN: 3261 ADR: 8,II |
|
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 103.0 to 107.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...