5'-Bromo-2'-hydroxypropiophenone ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
1-(5-Bromo-2-hydroxyphenyl)propan-1-one
B4769-1G
B4769-5G
TCIAB4769-1GEA
35.2
EUR
TCIAB4769-1G
TCIAB4769-5G
5'-Bromo-2'-hydroxypropiophenone ≥98.0% (by GC, titration analysis)
5'-Bromo-2'-hydroxypropiophenone
Formule:
C₉H₉BrO₂ Kookpunt: 156°C |
MDL Nummer:
MFCD00218577 CAS nummer: 17764-93-1 |
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 75.0 to 79.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...