N-(Benzoyloxy)succinimide ≥98.0% (by HPLC, total nitrogen)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
B4785-5G
B4785-25G
TCIAB4785-5GEA
85.8
EUR
TCIAB4785-5G
TCIAB4785-25G
N-(Benzoyloxy)succinimide ≥98.0% (by HPLC, total nitrogen)
N-(Benzoyloxy)succinimide
Formule:
C₁₁H₉NO₄ Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00078953 CAS nummer: 23405-15-4 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 135.0 to 139.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...