1-Benzhydrylazetidin-3-one ≥96.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

B4799-1G B4799-5G
TCIAB4799-1GEA 28.6 EUR
TCIAB4799-1G TCIAB4799-5G
1-Benzhydrylazetidin-3-one ≥96.0% (by GC, titration analysis)
1-Benzhydrylazetidin-3-one
Formule: C₁₆H₁₅NO
MDL Nummer: MFCD00964539
CAS nummer: 40320-60-3

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 96.0 %
Purity (Nonaqueous Titration) min. 96.0 %
Melting point 76.0 to 81.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR