6-Bromochromone-2-carboxylic acid ≥95.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

B4802-1G B4802-5G
TCIAB4802-1GEA 34.1 EUR
TCIAB4802-1G TCIAB4802-5G
6-Bromochromone-2-carboxylic acid ≥95.0% (by GC, titration analysis)
6-Bromochromone-2-carboxylic acid
Formule: C₁₀H₅BrO₄
Smeltpunt: 276°C
MDL Nummer: MFCD01242619
CAS nummer: 51484-06-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=619b98339731759aec7f14cdc3101952343731b7&vid=619b98339731759aec7f14cdc3101952343731b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR