2-Bromo-6-iodopyridine ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
B4815-1G B4815-5G
TCIAB4815-1GEA 77 EUR
TCIAB4815-1G TCIAB4815-5G
2-Bromo-6-iodopyridine ≥98.0% (by GC)
2-Bromo-6-iodopyridine
Formule: C₅H₃BrIN
MDL Nummer: MFCD08059557
CAS nummer: 234111-08-1
Order Now
Specification Test Results
Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 140.0 to 144.0 deg-C