1-Bromo-3,5-dihexylbenzene ≥96.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
B4818-1G
B4818-5G
TCIAB4818-1GEA
48.4
EUR
TCIAB4818-1G
TCIAB4818-5G
1-Bromo-3,5-dihexylbenzene ≥96.0% (by GC)
1-Bromo-3,5-dihexylbenzene
Formule:
C₁₈H₂₉Br |
CAS nummer:
1238156-36-9 |
Specification Test Results
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...