5-Bromo-4-chloro-3-indolyl acetate ≥98.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

B4830-50MG B4830-200MG
TCIAB4830-50MGEA 47.3 EUR
TCIAB4830-50MG TCIAB4830-200MG
5-Bromo-4-chloro-3-indolyl acetate ≥98.0% (by HPLC)
5-Bromo-4-chloro-3-indolyl acetate
Formule: C₁₀H₇BrClNO₂
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00037932
CAS nummer: 3252-36-6

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR