4-tert-Butylphenyl glycidyl ether ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

B4831-100G B4831-25G
TCIAB4831-100GEA 284 EUR
TCIAB4831-100G TCIAB4831-25G
4-tert-Butylphenyl glycidyl ether ≥98.0% (by GC)
4-tert-Butylphenyl glycidyl ether
Formule: C₁₃H₁₈O₂
Kookpunt: 294°C
Vlampunt: 102°C
MDL Nummer: MFCD00005136
CAS nummer: 3101-60-8
EINECS: 221-453-2

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR