2-Bromobenzimidazole ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 2-Bromo-1H-benzimidazole

B4838-1G B4838-5G
TCIAB4838-1GEA 51.7 EUR
TCIAB4838-1G TCIAB4838-5G
2-Bromobenzimidazole ≥98.0% (by GC, titration analysis)
2-Bromobenzimidazole
Formule: C₇H₅BrN₂
Smeltpunt: 192°C
MDL Nummer: MFCD00160009
CAS nummer: 54624-57-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR