4-Bromo-2-(trimethylsilyl)thiophene ≥97.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
B4856-200MG B4856-1G
TCIAB4856-200MGEA 99 EUR
TCIAB4856-200MG TCIAB4856-1G
4-Bromo-2-(trimethylsilyl)thiophene ≥97.0% (by GC)
4-Bromo-2-(trimethylsilyl)thiophene
Formule: C₇H₁₁BrSSi
Bewaar temperatuur: Koelkast
CAS nummer: 77998-61-9
Order Now
Specification Test Results
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %