2-Bromo-4-nitroaniline ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
B4873-1G
B4873-5G
TCIAB4873-1GEA
49.5
EUR
TCIAB4873-1G
TCIAB4873-5G
2-Bromo-4-nitroaniline ≥98.0% (by GC)
2-Bromo-4-nitroaniline
Formule:
C₆H₅BrN₂O₂ MW: 217.02 g/mol |
MDL Nummer:
MFCD00025152 CAS nummer: 13296-94-1 EINECS: 236-318-3 |
|
Specification Test Results
Appearance | White to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 104.0 to 108.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...