7,7'-Bi[1,4]benzoxazino[2,3,4-kl]phenoxazine ≥98.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: HN-D1

B4908-200MG B4908-1G
TCIAB4908-200MGEA 154 EUR
TCIAB4908-200MG TCIAB4908-1G
7,7'-Bi[1,4]benzoxazino[2,3,4-kl]phenoxazine ≥98.0% (by HPLC)
7,7'-Bi[1,4]benzoxazino[2,3,4-kl]phenoxazine
Formule: C₃₆H₂₀N₂O₄
MW: 544.57 g/mol
Smeltpunt: 376°C
CAS nummer: 1395881-55-6

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 4.80 to 5.50 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR