2,6-Bis(9H-carbazol-9-yl)pyridine ≥98.0% (by HPLC, total nitrogen)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

B4961-200MG B4961-1G
TCIAB4961-200MGEA 116 EUR
TCIAB4961-200MG TCIAB4961-1G
2,6-Bis(9H-carbazol-9-yl)pyridine ≥98.0% (by HPLC, total nitrogen)
2,6-Bis(9H-carbazol-9-yl)pyridine
Formule: C₂₉H₁₉N₃
Smeltpunt: 202°C
MDL Nummer: MFCD28138085
CAS nummer: 168127-49-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d578ec7bc392ff3198a894856bf8ee10799c1c20&vid=d578ec7bc392ff3198a894856bf8ee10799c1c20&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR