1-Butyl-2-thiourea ≥98.0% (by total nitrogen basis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

B4971-1G B4971-5G
TCIAB4971-1GEA 57.2 EUR
TCIAB4971-1G TCIAB4971-5G
1-Butyl-2-thiourea ≥98.0% (by total nitrogen basis)
1-Butyl-2-thiourea
Formule: C₅H₁₂N₂S
MDL Nummer: MFCD00022173
CAS nummer: 1516-32-1
EINECS: 216-165-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8133733b593f6588346badc0a9f3f1ee1a17b3e9&vid=8133733b593f6588346badc0a9f3f1ee1a17b3e9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Melting point 75.0 to 79.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR