N-(5-Bromopentyl)phthalimide ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
B4986-5G
B4986-25G
TCIAB4986-5GEA
85.8
EUR
TCIAB4986-5G
TCIAB4986-25G
N-(5-Bromopentyl)phthalimide ≥98.0% (by GC)
N-(5-Bromopentyl)phthalimide
Formule:
C₁₃H₁₄BrNO₂ MW: 296.16 g/mol |
MDL Nummer:
MFCD00060522 CAS nummer: 954-81-4 |
|
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 60.0 to 64.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...