N-(4-Biphenylyl)-1-naphthylamine ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: N-([1,1'-Biphenyl]-4-yl)naphthalen-1-amine

B5047-200MG B5047-1G
TCIAB5047-200MGEA 57.2 EUR
TCIAB5047-200MG TCIAB5047-1G
N-(4-Biphenylyl)-1-naphthylamine ≥98.0% (by GC)
N-(4-Biphenylyl)-1-naphthylamine
Formule: C₂₂H₁₇N
Kookpunt: 315°C
MDL Nummer: MFCD28138082
CAS nummer: 446242-37-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=069cb6364101f6efa01fab94cd0861a3c99436db&vid=069cb6364101f6efa01fab94cd0861a3c99436db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 147.0 to 151.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR