7-Bromoisoquinoline ≥96.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

B5071-200MG B5071-1G
TCIAB5071-200MGEA 51.7 EUR
TCIAB5071-200MG TCIAB5071-1G
7-Bromoisoquinoline ≥96.0% (by GC)
7-Bromoisoquinoline
Formule: C₉H₆BrN
MDL Nummer: MFCD07368661
CAS nummer: 58794-09-5

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 96.0 %
Melting point 69.0 to 73.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR