2-(3-Bromophenyl)pyridine ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

B5113-1G B5113-5G
TCIAB5113-1GEA 67.1 EUR
TCIAB5113-1G TCIAB5113-5G
2-(3-Bromophenyl)pyridine ≥98.0% (by GC)
2-(3-Bromophenyl)pyridine
Formule: C₁₁H₈BrN
MDL Nummer: MFCD09749846
CAS nummer: 4373-60-8

Order Now

Specification Test Results

Appearance White to Yellow to Green clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR