Boc-Abu-OH ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: (S)-2-(tert-Butoxycarbonylamino)butyric acid, Boc-L-2-aminobutanoic acid, BOC-L-α-Aminobutyric acid, Boc-L-Abu-OH, (S)-2-(Boc-amino)butyric acid

B5115-5G B5115-25G
TCIAB5115-5GEA 18.7 EUR
TCIAB5115-5G TCIAB5115-25G
Boc-Abu-OH ≥98.0% (by HPLC, titration analysis)
Boc-Abu-OH
Formule: C₉H₁₇NO₄
MW: 203.238 g/mol
MDL Nummer: MFCD00037267
CAS nummer: 34306-42-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=40d24e688ea1fae50da20c4e27c9617fbedc9ff4&vid=5e9666d32b1bf3896159f9000adb2720586d10eb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 73.0 to 77.0 deg-C
Specific rotation [a]20/D -17.0 to -20.0 deg (C=1,MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR