Bosentan monohydrate ≥98.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 4-tert-Butyl-N-[6-(2-hydroxyethoxy)-5-(2-methoxyphenoxy)-2-(pyrimidin-2-yl)pyrimidin-4-yl]benzenesulphonamide, 4-(tert-Butyl)-N-(6-(2-hydroxyethoxy)-5-(2-methoxyphenoxy)-[2,2'-bipyrimidin]-4-yl)benzenesulphonamide hydrate, Bosentan hydrate, 4-(tert-Butyl)-N-[6-(2-hydroxyethoxy)-5-(2-methoxyphenoxy)[2,2'-bipyrimidin]-4-yl]benzenesulphonamide

B5118-25MG B5118-100MG
TCIAB5118-25MGEA 92.4 EUR
TCIAB5118-25MG TCIAB5118-100MG
Bosentan monohydrate ≥98.0% (by HPLC)
Bosentan monohydrate
Formule: C₂₇H₂₉N₅O₆S·H₂O
MDL Nummer: MFCD09751188
CAS nummer: 157212-55-0
Merck Index: 14,01353
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39f03aef0e3ec9059f3d1be7bd14fb8a701ebdfe&vid=39f03aef0e3ec9059f3d1be7bd14fb8a701ebdfe&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 11.60 to 12.90 %
Water 2.5 to 4.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR