2-Bromo-9,9-dibutylfluorene ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
B5127-1G
B5127-5G
TCIAB5127-1GEA
74.8
EUR
TCIAB5127-1G
TCIAB5127-5G
2-Bromo-9,9-dibutylfluorene ≥98.0% (by GC)
2-Bromo-9,9-dibutylfluorene
Formule:
C₂₁H₂₅Br |
MDL Nummer:
MFCD12031333 CAS nummer: 88223-35-2 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 53.0 to 57.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...