4-Bromo-1,2,3,6,7,8-hexahydropyrene ≥96.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

B5129-1G B5129-5G
TCIAB5129-1GEA 330 EUR
TCIAB5129-1G TCIAB5129-5G
4-Bromo-1,2,3,6,7,8-hexahydropyrene ≥96.0% (by GC)
4-Bromo-1,2,3,6,7,8-hexahydropyrene
Formule: C₁₆H₁₅Br
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD01571021
CAS nummer: 1732-25-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f1da2646941c6e0a31dce996d378981308f821cb&vid=f1da2646941c6e0a31dce996d378981308f821cb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %
Melting point 127.0 to 131.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR